Influence of harvesting technique and maceration process on aroma and phenolic attributes of Sauvignon blanc wine.
Food chemistry (2015-04-13)
Kenneth J Olejar, Bruno Fedrizzi, Paul A Kilmartin
RESUMEN
Sauvignon blanc wines are characterised by their varietal aromas and low phenolic content. Mechanical harvesting has been shown to increase several varietal aromas. Likewise, maceration techniques have produced increases in phenolic content and antioxidant activity, but these can also alter tactile attributes and sensory profiles. Mechanical harvesting and cryogenic maceration were used in combination to produce a Sauvignon blanc wine with increased phenolic content and antioxidant activity, while showing a similar sensory profile to control wines. Phenolic profiles of the wines showed differences between the harvesting and maceration techniques. Mechanical harvesting contributed to decreases in phenolics through reaction with oxidative radicals. Cryogenic maceration increased phenolics and antioxidant activity. Cryogenic maceration also increased the levels of several varietal aromas, for Sauvignon blanc wines made from both hand-picked and from machine-harvested fruit. Furthermore, cryogenic treatment of hand-picked fruit increased varietal thiols to levels similar to machine-harvested control wines.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)