Fucoxanthin increases lifespan of Drosophila melanogaster and Caenorhabditis elegans.
Pharmacological research (2015-08-21)
Ekaterina Lashmanova, Ekaterina Proshkina, Svetlana Zhikrivetskaya, Oksana Shevchenko, Elena Marusich, Sergey Leonov, Alex Melerzanov, Alex Zhavoronkov, Alexey Moskalev
RESUMEN
The pharmacological activation of stress-defense mechanisms is one of the perspective ways to increase human lifespan. The goal of the present study was to study the effects on lifespan of Drosophila melanogaster and Caenorhabditis elegans of two carotenoids: ß-carotene and fucoxanthin, which are bioactive natural substances in human diet. In addition, the effects of carotenoids on the flies survival were studied under stress conditions, including starvation, thermal stress (35°C), oxidative stress (20 mM paraquat), as well as locomotor activity, fecundity, and genes expression level. Our results demonstrated lifespan extension of flies by both carotenoids. However, the positive effects on the lifespan of C. elegans were revealed only for fucoxanthin. In presence of carotenoids decreased flies' fecundity, increased spontaneous locomotor activity and resistance to oxidative stress were detected.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Fucoxanthin, carotenoid antioxidant