Phosphorus migration analysis using synchrotron radiation in soil treated with Brazilian granular fertilizers.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2015-09-08)
Robson C de Castro, Vinícius de Melo Benites, Paulo César Teixeira, Marcelino José Dos Anjos, Luis Fernando de Oliveira
RESUMEN
The aim of this study was to evaluate the phosphorus (P) mobility in a tropical Brazilian soil type red Oxisol treated with three different forms of granular fertilizer. Total Reflection X-Ray Fluorescence (TXRF) was applied to determine the concentration of P at different distances from granular fertilizer application point. The results showed that most of the P from fertilizers tends to concentrate in a region of up to 10mm around the place of the fertilizer deposition.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%