  • Search Results
  • A Positive Allosteric Modulator of the Adenosine A1 Receptor Selectively Inhibits Primary Afferent Synaptic Transmission in a Neuropathic Pain Model.
A Positive Allosteric Modulator of the Adenosine A1 Receptor Selectively Inhibits Primary Afferent Synaptic Transmission in a Neuropathic Pain Model.
Molecular pharmacology (2015-06-25)
Wendy L Imlach, Rebecca F Bhola, Lauren T May, Arthur Christopoulos, Macdonald J Christie
ABSTRACT
In the spinal cord and periphery, adenosine inhibits neuronal activity through activation of the adenosine A1 receptor (A1R), resulting in antinociception and highlighting the potential of therapeutically targeting the receptor in the treatment of neuropathic pain. This study investigated the changes in adenosine tone and A1R signaling, together with the actions of a novel A1R positive allosteric modulator (PAM), VCP171 [(2-amino-4-(3-(trifluoromethyl)phenyl)thiophen-3-yl)(phenyl)methanone], on excitatory and inhibitory neurotransmission at spinal cord superficial dorsal horn synapses in a rat partial nerve-injury model of neuropathic pain. In the absence of A1R agonists, superfusion of the A1R antagonist, 8-cyclopentyl-1,3-dipropylxanthine (DPCPX; 1 μM), produced a significantly greater increase in electrically evoked α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor-mediated synaptic current (eEPSC) amplitude in both lamina I and II neurons from nerve-injured animals than in controls, suggesting that endogenous adenosine tone is increased in the dorsal horn. Inhibitory GABAergic and glycinergic synaptic currents were also significantly increased by DPCPX in controls but there was no difference after nerve injury. The A1R agonist, N6-cyclopentyladenosine, produced greater inhibition of eEPSC amplitude in lamina II but not lamina I of the spinal cord dorsal horn in nerve-injured versus control animals, suggesting a functional increase in A1R sensitivity in lamina II neurons after nerve injury. The A1R PAM, VCP171, produced a greater inhibition of eEPSC amplitude of nerve-injury versus control animals in both lamina I and lamina II neurons. Enhanced adenosine tone and A1R sensitivity at excitatory synapses in the dorsal horn after nerve injury suggest that new generation PAMs of the A1R can be effective treatments for neuropathic pain.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture