Merck
  • Inicio
  • Resultados de la búsqueda
  • Making sense of nickel accumulation and sub-lethal toxic effects in saline waters: Fate and effects of nickel in the green crab, Carcinus maenas.
Making sense of nickel accumulation and sub-lethal toxic effects in saline waters: Fate and effects of nickel in the green crab, Carcinus maenas.
Aquatic toxicology (Amsterdam, Netherlands) (2015-04-29)
Tamzin A Blewett, Chris N Glover, Sandra Fehsenfeld, Michael J Lawrence, Som Niyogi, Greg G Goss, Chris M Wood
RESUMEN
In freshwater, invertebrates nickel (Ni) is considered an ionoregulatory toxicant, but its mechanism of toxicity in marine settings, and how this varies with salinity, is poorly understood. This study investigated Ni accumulation and physiological mechanisms of sub-lethal Ni toxicity in the euryhaline green crab Carcinus maenas. Male crabs were exposed to 8.2μg/L (the US EPA chronic criterion concentration for salt waters) of waterborne Ni (radiolabelled with (63)Ni) at three different salinities, 20%, 60% and 100% SW for 24h. Whole body Ni accumulation in 20% SW was 3-5 fold greater than in 60% or 100% SW, and >80% of accumulated Ni was in the carapace at all salinities. Ni also accumulated in posterior gill 8, which showed a higher accumulation in 20% SW than in other salinities, a pattern also seen at higher exposure concentrations of Ni (500 and 3000μg/L). Gill perfusion experiments revealed that Ni was taken up by both anterior and posterior gills, but in 20% SW the posterior gill 8, which performs ionoregulatory functions, accumulated more Ni than the anterior gill 5, which primarily has a respiratory function. The sub-lethal consequences of Ni exposure were investigated by placing crabs in Ni concentrations of 8.2, 500, and 3000μg/L at 20, 60 or 100% SW for 24h. In 20% SW, haemolymph Ca levels were significantly decreased by exposure to Ni concentrations of 8.2μg/L or higher, whereas Na concentrations were depressed only at 3000μg/L. Na(+)/K(+)-ATPase activity was inhibited at both 500 and 3000μg/L in gill 8, but only in 20% SW. Haemolymph K, Mg, and osmolality were unaffected throughout, though all varied with salinity in the expected fashion. These data suggest that Ni impacts ionoregulatory function in the green crab, in a gill- and salinity-dependent manner.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)