Hydroxylammonium Tetrafluoridooxidovanadate(V) - (NH3OH)[VOF4].
Acta chimica Slovenica (2015-06-19)
Matic Lozinšek
RESUMEN
Hydroxylammonium tetrafluoridooxidovanadate(V) has been prepared by reaction of (NH3OH)Cl and VOF3 in anhydrous HF solvent and characterised in the solid state by single-crystal X-ray diffraction and Raman spectroscopy. Colourless (NH3OH)[VOF4] crystallises in the monoclinic P21/n space group with a = 7.2349(2) Å, b = 5.0351(2) Å, c = 12.8322(4) Å, β = 94.843(3)°, V = 465.79(3) Å3 and Z = 4 at 150 K. The crystal structure of the title compound is comprised of polymeric chain anions, [VOF4]nn-, interconnected by hydrogen bonds with hydroxylammonium cations, (NH3OH)+.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Perfluorodecalin, 95%
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%