  • Search Results
  • Enantiomer separation of acidic chiral compounds on a quinine-silica/zirconia hybrid monolith by capillary electrochromatography.
Enantiomer separation of acidic chiral compounds on a quinine-silica/zirconia hybrid monolith by capillary electrochromatography.
Journal of chromatography. A (2015-04-22)
Le Ngoc Tran, Jung Hag Park
ABSTRACT
A weak anion-exchanger chiral selector, quinine-incorporated silica/zirconia hybrid monolithic (QUI-S/ZHM) capillary column was prepared by sol-gel technology. The performance of the QUI-S/ZHM column was investigated for enantioresolution of a set of acidic chiral drugs and dinitrobenzoyl (DNB)-amino acids by capillary electrochromatography in aqueous organic mobile phases composed of acetonitrile (ACN) and triethylammonium acetate (TEAA) buffer. Effects of several parameters including the ACN content, concentration and pH of the mobile phase on the chiral separation were examined. Baseline resolutions of all the compounds were obtained in the mobile phase consisting of 70:30 ACN/TEAA (10mM, pH 6) under applied voltage of -10kV at 25°C within 20min.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
3-(Triethoxysilyl)propyl isocyanate, 95%