Merck
  • Inicio
  • Resultados de la búsqueda
  • Improved sensitivity by use of gas chromatography-positive chemical ionization triple quadrupole mass spectrometry for the analysis of drug related substances.
Improved sensitivity by use of gas chromatography-positive chemical ionization triple quadrupole mass spectrometry for the analysis of drug related substances.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-08-22)
Wim Van Gansbeke, Michael Polet, Fiona Hooghe, Christophe Devos, Peter Van Eenoo
RESUMEN
In 2013, the World Anti-Doping Agency (WADA) drastically lowered the minimum required performance levels (MRPLs) of most doping substances, demanding a substantial increase in sensitivity of the existing methods. For a number of compounds, conventional electron impact ionization gas chromatography tandem mass spectrometry (GC-EI-MS/MS) is often no longer sufficient to reach these MRPLs and new strategies are required. In this study, the capabilities of positive ion chemical ionization (PICI) GC-MS/MS are investigated for a wide range of drug related compounds of various classes by injection of silylated reference standards. Ammonia as PICI reagent gas had superior characteristics for GC-MS/MS purposes than methane. Compared to GC-EI-MS/MS, PICI (with ammonia as reagent gas) provided more selective ion transitions and consequently, increased sensitivity by an average factor of 50. The maximum increase (by factor of 500-1000) was observed in the analysis of stimulants, namely chlorprenaline, furfenorex and phentermine. In total, improved sensitivity was obtained for 113 out of 120 compounds. A new GC-PICI-MS/MS method has been developed and evaluated for the detection of a wide variety of exogenous doping substances and the quantification of endogenous steroids in urine in compliance with the required MRPLs established by WADA in 2013. The method consists of a hydrolysis and extraction step, followed by derivatization and subsequent 1μL pulsed splitless injection on GC-PICI-MS/MS (16min run). The increased sensitivity allows the set up of a balanced screening method that meets the requirements for both quantitative and qualitative compounds: sufficient capacity and resolution in combination with high sensitivity and short analysis time. This resulted in calibration curves with a wide linear range (e.g., 48-9600ng/mL for androsterone and etiochanolone; all r(2)>0.99) without compromising the requirements for the qualitative compounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ethanethiol, 97%
Sigma-Aldrich
Ammonium iodide, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Ethanethiol, ≥97%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis