  • Search Results
  • Weight gain and inflammation regulate aromatase expression in male adipose tissue, as evidenced by reporter gene activity.
Weight gain and inflammation regulate aromatase expression in male adipose tissue, as evidenced by reporter gene activity.
Molecular and cellular endocrinology (2015-06-10)
L Polari, E Yatkin, M G Martínez Chacón, M Ahotupa, A Smeds, L Strauss, F Zhang, M Poutanen, N Saarinen, S I Mäkelä
ABSTRACT
Obesity and white adipose tissue (WAT) inflammation are associated with enhanced aromatization in women, but little is known about the regulation of aromatase (CYP19A1) gene expression in male WAT. We investigated the impact of weight gain and WAT inflammation on the regulation of CYP19A1 in males, by utilizing the hARO-Luc aromatase reporter mouse model containing a >100-kb 5'-region of the human CYP19A1 gene. We show that hARO-Luc reporter activity is enhanced in WAT of mice with increased adiposity and inflammation. Dexamethasone and TNFα, as well as forskolin and phorbol 12-myristate 13-acetate, upregulate hARO-Luc activity, suggesting the involvement of promoters I.4 and I.3/II. Furthermore, we show that diet enriched with antioxidative plant polyphenols attenuates WAT inflammation and hARO-Luc activity in obese males. In conclusion, our data suggest that obesity-associated WAT inflammation leads to increased peripheral CYP19A1 expression in males, and that polyphenol-enriched diet may have the potential to attenuate excessive aromatization in WAT of obese men.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%