• Inicio
  • Resultados de la búsqueda
  • Human Chorionic Gonadotropin Protects Vascular Endothelial Cells from Oxidative Stress by Apoptosis Inhibition, Cell Survival Signalling Activation and Mitochondrial Function Protection.
Human Chorionic Gonadotropin Protects Vascular Endothelial Cells from Oxidative Stress by Apoptosis Inhibition, Cell Survival Signalling Activation and Mitochondrial Function Protection.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-08-19)
Daniela Surico, Serena Farruggio, Patrizia Marotta, Giulia Raina, David Mary, Nicola Surico, Giovanni Vacca, Elena Grossini
RESUMEN
Previous reports have made it hypothetically possible that human chorionic gonadotropin (hCG) could protect against the onset of pregnancy-related pathological conditions by acting as an antioxidant. In the present study we planned to examine the effects of hCG against oxidative stress in human umbilical vein endothelial cells (HUVEC). HUVEC were subjected to peroxidation by hydrogen peroxide. The modulation of nitric oxide (NO) release by hCG and its effects on cell viability, glutathione (GSH) levels, mitochondrial membrane potential and mitochondrial transition pore opening (MPTP) were examined by specific dyes. Endothelial and inducible NO synthase (eNOS and iNOS), Akt and extracellular -signal-regulated kinases 1/2 (ERK1/2) activation and markers of apoptosis were analyzed by Western Blot. In HUVEC, hCG reduced NO release by modulating eNOS and iNOS. Moreover, hCG protected HUVEC against oxidative stress by preventing GSH reduction and apoptosis, by maintaining Akt and ERK1/2 activation and by keeping mitochondrial function. The present results have for the first time shown protective effects exerted by hCG on vascular endothelial function, which would be achieved by modulation of NO release, antioxidant and antiapoptotic actions and activation of cell survival signalling. These findings could have clinical implications in the management of pregnancy-related disorders.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets