Development and validation of an LC-MS/MS method for determination of p-phenylenediamine and its metabolites in blood samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-06-16)
Khaled M Mohamed, Duncan Cromarty, Vanessa Steenkamp
RESUMEN
In some developing countries, p-phenylenediamine (PPD) is used in combination with Henna as hair dye or skin decoration. A sensitive LC-MS/MS method was developed and validated for the simultaneous determination of p-phenylenediamine (PPD) and its metabolites N-acetyl-p-phenylenediamine (MAPPD) and N,N-diacetyl-p-phenylenediamine (DAPPD) in human blood. Acetanilide was used as an internal standard (IS). The LC-MS/MS was operated under multiple reaction-monitoring mode using the electrospray positive ionization technique. The transition ions m/z 109→92, m/z 151→92, m/z 193→92, and m/z 136→77 were selected for the quantification of PPD, MAPPD, DAPPD, and IS, respectively. The linear range was 10-2000ng/mL for all the compounds. The absolute recoveries were 51.94, 56.20 and 54.88% for PPD, MAPPD and DAPPD, respectively. Intra- and inter-assay imprecision were lower than 14% (RSD), and the bias of the assay was lower than 15% for all the compounds. The stability studies demonstrated that critical degradation for PPD in blood samples and autosampler occurred after 6h, while MAPPD and DAPPD were stable in blood samples and the autosampler up to 48h and 24h, respectively. This newly developed method allows for the detection of PPD and its metabolites in blood samples in the clinical and forensic setting.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Acetanilide, 99%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetanilide, purified by sublimation, ≥99.9%
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
1,4-Phenylenediamine dihydrochloride, ≥99.0%
Sigma-Aldrich
Acetanilide, zone-refined, purified by sublimation, ≥99.95%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
Sigma-Aldrich
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)