Merck
  • Inicio
  • Resultados de la búsqueda
  • A cell-based screening for TAZ activators identifies ethacridine, a widely used antiseptic and abortifacient, as a compound that promotes dephosphorylation of TAZ and inhibits adipogenesis in C3H10T1/2 cells.
A cell-based screening for TAZ activators identifies ethacridine, a widely used antiseptic and abortifacient, as a compound that promotes dephosphorylation of TAZ and inhibits adipogenesis in C3H10T1/2 cells.
Journal of biochemistry (2015-05-17)
Shodai Kawano, Junichi Maruyama, Shunta Nagashima, Kazutoshi Inami, Wenzhe Qiu, Hiroaki Iwasa, Kentaro Nakagawa, Mari Ishigami-Yuasa, Hiroyuki Kagechika, Hiroshi Nishina, Yutaka Hata
RESUMEN
Transcriptional co-activator with PSD-95/Dlg-A/ZO-1 (PDZ)-binding motif (TAZ) regulates in cell proliferation and differentiation. In mesenchymal stem cells it promotes osteogenesis and myogenesis, and suppresses adipogenesis. TAZ activators are expected to prevent osteoporosis, obesity and muscle atrophy. TAZ activation induces epithelial-mesenchymal transition, confers stemness to cancer cells and leads to poor clinical prognosis in cancer patients. In this point of view, TAZ inhibitors should contribute to cancer therapy. Thus, TAZ attracts attention as a two-faced drug target. We screened for TAZ modulators by using human lung cancer A549 cells expressing the fluorescent reporter. Through this assay, we obtained TAZ activator candidates. We unexpectedly found that ethacridine, a widely used antiseptic and abortifacient, enhances the interaction of TAZ and protein phosphatases and increases unphosphorylated and nuclear TAZ. Ethacridine inhibits adipogenesis in mesenchymal C3H10T1/2 cells through the activation of TAZ. This finding suggests that ethacridine is a bona fide TAZ activator and supports that our assay is useful to discover TAZ activators.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M