Near-infrared fluorescent digital pathology for the automation of disease diagnosis and biomarker assessment.
Molecular imaging (2015-03-31)
Summer L Gibbs, Elizabeth Genega, Jeffery Salemi, Vida Kianzad, Haley L Goodwill, Yang Xie, Rafiou Oketokoun, Parmeshwar Khurd, Ali Kamen, John V Frangioni
RESUMEN
Hematoxylin-eosin (H&E) staining of tissue has been the mainstay of pathology for more than a century. However, the learning curve for H&E tissue interpretation is long, whereas intra- and interobserver variability remain high. Computer-assisted image analysis of H&E sections holds promise for increased throughput and decreased variability but has yet to demonstrate significant improvement in diagnostic accuracy. Addition of biomarkers to H&E staining can improve diagnostic accuracy; however, coregistration of immunohistochemical staining with H&E is problematic as immunostaining is completed on slides that are at best 4 μm apart. Simultaneous H&E and immunostaining would alleviate coregistration problems; however, current opaque pigments used for immunostaining obscure H&E. In this study, we demonstrate that diagnostic information provided by two or more independent wavelengths of near-infrared (NIR) fluorescence leave the H&E stain unchanged while enabling computer-assisted diagnosis and assessment of human disease. Using prostate cancer as a model system, we introduce NIR digital pathology and demonstrate its utility along the spectrum from prostate biopsy to whole mount analysis of H&E-stained tissue.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
Xylenes, reagent grade
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Supelco
Hematoxylin
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Eosin Y
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder