Multiple inputs control sulfur-containing amino acid synthesis in Saccharomyces cerevisiae.
Molecular biology of the cell (2014-03-22)
Meru J Sadhu, James J Moresco, Anjali D Zimmer, John R Yates, Jasper Rine
RESUMEN
In Saccharomyces cerevisiae, transcription of the MET regulon, which encodes the proteins involved in the synthesis of the sulfur-containing amino acids methionine and cysteine, is repressed by the presence of either methionine or cysteine in the environment. This repression is accomplished by ubiquitination of the transcription factor Met4, which is carried out by the SCF(Met30) E3 ubiquitin ligase. Mutants defective in MET regulon repression reveal that loss of Cho2, which is required for the methylation of phosphatidylethanolamine to produce phosphatidylcholine, leads to induction of the MET regulon. This induction is due to reduced cysteine synthesis caused by the Cho2 defects, uncovering an important link between phospholipid synthesis and cysteine synthesis. Antimorphic mutants in S-adenosyl-methionine (SAM) synthetase genes also induce the MET regulon. This effect is due, at least in part, to SAM deficiency controlling the MET regulon independently of SAM's contribution to cysteine synthesis. Finally, the Met30 protein is found in two distinct forms whose relative abundance is controlled by the availability of sulfur-containing amino acids. This modification could be involved in the nutritional control of SCF(Met30) activity toward Met4.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid