  • Search Results
  • Altered expression of uncoupling protein 2 in GLP-1-producing cells after chronic high glucose exposure: implications for the pathogenesis of diabetes mellitus.
Altered expression of uncoupling protein 2 in GLP-1-producing cells after chronic high glucose exposure: implications for the pathogenesis of diabetes mellitus.
American journal of physiology. Cell physiology (2016-01-08)
Francesca Urbano, Agnese Filippello, Antonino Di Pino, Davide Barbagallo, Stefania Di Mauro, Alessandro Pappalardo, Agata Maria Rabuazzo, Michele Purrello, Francesco Purrello, Salvatore Piro
ABSTRACT
Glucagon-like peptide-1 (GLP-1) is a gut L-cell hormone that enhances glucose-stimulated insulin secretion. Several approaches that prevent GLP-1 degradation or activate the GLP-1 receptor are being used to treat type 2 diabetes mellitus (T2DM) patients. In T2DM, GLP-1 secretion has been suggested to be impaired, and this defect appears to be a consequence rather than a cause of impaired glucose homeostasis. However, although defective GLP-1 secretion has been correlated with insulin resistance, little is known about the direct effects of chronic high glucose concentrations, which are typical in diabetes patients, on GLP-1-secreting cell function. In the present study, we demonstrate that glucotoxicity directly affects GLP-1 secretion in GLUTag cells chronically exposed to high glucose. Our results indicate that this abnormality is associated with a decrease in ATP production due to the elevated expression of mitochondrial uncoupling protein 2 (UCP2). Furthermore, UCP2 inhibition using small interfering RNA (siRNA) and the application of glibenclamide, an ATP-sensitive potassium (KATP(+)) channel blocker, reverse the GLP-1 secretion defect induced by chronic high-glucose treatment. These results show that glucotoxicity diminishes the secretory responsiveness of GLP-1-secreting cells to acute glucose stimulation. We conclude that the loss of the incretin effect, as observed in T2DM patients, could at least partially depend on hyperglycemia, which is typical in diabetes patients. Such an understanding may not only provide new insight into diabetes complications but also ultimately contribute to the identification of novel molecular targets within intestinal L-cells for controlling and improving endogenous GLP-1 secretion.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Ile-Pro-Ile, ≥97% (HPLC)