  • Search Results
  • Targeting peroxiredoxin I potentiates 1,25-dihydroxyvitamin D3-induced cell differentiation in leukemia cells.
Targeting peroxiredoxin I potentiates 1,25-dihydroxyvitamin D3-induced cell differentiation in leukemia cells.
Molecular medicine reports (2016-01-20)
Wei Wei, Chuanxu Liu, Dongjun Qin, Lili Song, Li Xia, Hu Lei, Yun Yu, Weiwei Wang, Jianxin Pu, Handong Sun, Yingli Wu, Hanzhang Xu, Siguo Hao
ABSTRACT
Although 1,25‑dihydroxyvitamin D3 (VD3) is regarded as a promising inducing agent for leukemia cell differentiation, it is not as effective an agent as all‑trans‑retinoic acid, and its usefulness is also limited by the adverse effects of hypercalcemia. The aim of the present study was to determine whether combining VD3 with adenanthin, a peroxiresoxin I (Prx I)‑targeting natural compound, improves the efficacy of VD3. Cell viability was assessed using a trypan blue exclusion assay and flow cytometry was used to evaluate the expression of cell surface markers, CD11b/CD14, and the level of reactive oxygen species (ROS). Wright's staining was used to examine morphological changes and RNA‑interference was used to knockdown Prx I and p65 gene expression. Protein expression was determined by western blot analysis. The results demonstrated that adenanthin markedly enhanced VD3‑induced cell differentiation of leukemia NB4 cells, as evidenced by the increased percentage of CD11b‑ and CD14‑positive cells, the mature morphology of the monocytes and the increased phagocytic ability. Consistent with these results, knockdown of Prx I, but not nuclear factor‑κB (p65), enhanced VD3‑induced cell differentiation. The combinatorial effects of adenanthin and VD3 were shown to be associated with the ROS‑CCAAT‑enhancer‑binding protein (C/EBP)β axis, since N‑acetylcysteine, a ROS scavenger, was able to abrogate the differentiation‑enhancing effects of adenanthin, and the knockdown of C/EBPβ also inhibited the combinatorial effects of adenanthin and VD3. In addition, co‑treatment with adenanthin and VD3 was able to induce differentiation in other non‑acute promyelocytic leukemia cells and primary leukemia cells. In conclusion, the results of the present study revealed a novel role for Prx I in VD3‑induced cell differentiation, and suggested that targeting Prx I may represent a novel strategy to enhance VD3‑induced leukemia cell differentiation.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorofluorescin diacetate, ≥97%