Low-dose laulimalide represents a novel molecular probe for investigating microtubule organization.
Cell cycle (Georgetown, Tex.) (2012-08-09)
Melissa J Bennett, Gordon K Chan, J B Rattner, David C Schriemer
RESUMEN
Laulimalide is a natural product that has strong taxoid-like properties but binds to a distinct site on β-tubulin in the microtubule (MT) lattice. At elevated concentrations, it generates MTs that are resistant to depolymerization, and it induces a conformational state indistinguishable from taxoid-treated MTs. In this study, we describe the effect of low-dose laulimalide on various stages of the cell cycle and compare these effects to docetaxel as a representative of taxoid stabilizers. No evidence of MT bundling in interphase was observed with laulimalide, in spite of the fact that MTs are stabilized at low dose. Cells treated with laulimalide enter mitosis but arrest at prometaphase by generating multiple asters that coalesce into supernumerary poles and interfere with the integrity of the metaphase plate. Cells with a preformed bipolar spindle exist under heightened tension under laulimalide treatment, and chromosomes rapidly shear from the plate, even though the bipolar spindle is well-preserved. Docetaxel generates a similar phenotype for HeLa cells entering mitosis, but when treated at metaphase, cells undergo chromosomal fragmentation and demonstrate reduced centromere dynamics, as expected for a taxoid. Our results suggest that laulimalide represents a new class of molecular probe for investigating MT-mediated events, such as kinetochore-MT interactions, which may reflect the location of the ligand binding site within the interprotofilament groove.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.