Chk2-dependent phosphorylation of XRCC1 in the DNA damage response promotes base excision repair.
The EMBO journal (2008-10-31)
Wen-Cheng Chou, Hui-Chun Wang, Fen-Hwa Wong, Shian-ling Ding, Pei-Ei Wu, Sheau-Yann Shieh, Chen-Yang Shen
RESUMEN
The DNA damage response (DDR) has an essential function in maintaining genomic stability. Ataxia telangiectasia-mutated (ATM)-checkpoint kinase 2 (Chk2) and ATM- and Rad3-related (ATR)-Chk1, triggered, respectively, by DNA double-strand breaks and blocked replication forks, are two major DDRs processing structurally complicated DNA damage. In contrast, damage repaired by base excision repair (BER) is structurally simple, but whether, and how, the DDR is involved in repairing this damage is unclear. Here, we demonstrated that ATM-Chk2 was activated in the early response to oxidative and alkylation damage, known to be repaired by BER. Furthermore, Chk2 formed a complex with XRCC1, the BER scaffold protein, and phosphorylated XRCC1 in vivo and in vitro at Thr(284). A mutated XRCC1 lacking Thr(284) phosphorylation was linked to increased accumulation of unrepaired BER intermediate, reduced DNA repair capacity, and higher sensitivity to alkylation damage. In addition, a phosphorylation-mimic form of XRCC1 showed increased interaction with glycosylases, but not other BER proteins. Our results are consistent with the phosphorylation of XRCC1 by ATM-Chk2 facilitating recruitment of downstream BER proteins to the initial damage recognition/excision step to promote BER.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid