Merck
  • Inicio
  • Resultados de la búsqueda
  • Effects of the antimicrobial contaminant triclocarban, and co-exposure with the androgen 17β-trenbolone, on reproductive function and ovarian transcriptome of the fathead minnow (Pimephales promelas).
Effects of the antimicrobial contaminant triclocarban, and co-exposure with the androgen 17β-trenbolone, on reproductive function and ovarian transcriptome of the fathead minnow (Pimephales promelas).
Environmental toxicology and chemistry (2016-06-18)
Daniel L Villeneuve, Kathleen M Jensen, Jenna E Cavallin, Elizabeth J Durhan, Natàlia Garcia-Reyero, Michael D Kahl, Richard L Leino, Elizabeth A Makynen, Leah C Wehmas, Edward J Perkins, Gerald T Ankley
RESUMEN
Triclocarban (TCC) is an antimicrobial agent routinely detected in surface waters that has been hypothesized to interact with the vertebrate endocrine system. The present study examined the effects of TCC alone and in combination with the model endocrine disruptor 17β-trenbolone (TRB) on fish reproductive function. Adult Pimephales promelas were continuously exposed to either 1 µg TCC/L or 5 µg TCC/L, to 0.5 µg TRB/L, or to a mixture (MIX) of 5 µg TCC/L and 0.5 µg TRB/L for 22 d, and a variety of reproductive and endocrine-related endpoints were examined. Cumulative fecundity was significantly reduced in fathead minnows exposed to TRB, MIX, or 5 µg TCC/L. Exposure to 1 µg TCC/L had no effect on reproduction. In general, both TRB and MIX treatments caused similar physiological effects, evoking significant reductions in female plasma vitellogenin, estradiol, and testosterone, and significant increases in male plasma estradiol. Based on analysis of the ovarian transcriptome, there were potential pathway impacts that were common to both TRB- and TCC-containing treatment groups. In most cases, however, those pathways were more plausibly linked to differences in reproductive status than to androgen-specific functions. Overall, TCC was reproductively toxic to fish at concentrations at or near those that have been measured in surface water. There was little evidence that TCC elicits reproductive toxicity through a specific mode of endocrine or reproductive action, nor that it could augment the androgenic effects of TRB. Nonetheless, the relatively small margin of safety between some measured environmental concentrations and effect concentrations suggests that concern is warranted. Environ Toxicol Chem 2017;36:231-242. Published 2016 Wiley Periodicals Inc. on behalf of SETAC. This article is a US government work and, as such, is in the public domain in the United States of America.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Medium 199, HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%