  • Search Results
  • TAp63 is important for cardiac differentiation of embryonic stem cells and heart development.
TAp63 is important for cardiac differentiation of embryonic stem cells and heart development.
Stem Cells (2011)
Rouleau M