MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Promiscuous dimerization of the growth hormone secretagogue receptor (GHS-R1a) attenuates ghrelin-mediated signaling.
Promiscuous dimerization of the growth hormone secretagogue receptor (GHS-R1a) attenuates ghrelin-mediated signaling.
The Journal of Biological Chemistry (2013)
Schellekens H et al.