MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Immunoassays for riboflavin and flavin mononucleotide using antibodies specific to d-ribitol and d-ribitol-5-phosphate.
Immunoassays for riboflavin and flavin mononucleotide using antibodies specific to d-ribitol and d-ribitol-5-phosphate.
Journal of immunological methods (2017-03-23)
G Ravi, Yeldur P Venkatesh
PMID
28327345
ABSTRACT
Riboflavin (vitamin B
Product Description
P5669
Sigma-Aldrich
Pyridoxine, ≥98%
Pricing
103217
Sigma-Aldrich
Lumichrome
Pricing
415413
Sigma-Aldrich
Sodium hydroxide solution, 50% in H
2
O
Pricing