A circular dichroism sensor for selective detection of Cd
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-05-06)
Pradthana Sianglam, Sirinan Kulchat, Thawatchai Tuntulani, Wittaya Ngeontae
RESUMEN
We demonstrate an advance in the fabrication of circular dichroism (CD) sensors for detection of Cd
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Penicillamine, 99%