TGFbeta induces SIK to negatively regulate type I receptor kinase signaling.
The Journal of cell biology (2008-08-30)
Marcin Kowanetz, Peter Lönn, Michael Vanlandewijck, Katarzyna Kowanetz, Carl-Henrik Heldin, Aristidis Moustakas
RESUMEN
Signal transduction by transforming growth factor beta (TGFbeta) coordinates physiological responses in diverse cell types. TGFbeta signals via type I and type II receptor serine/threonine kinases and intracellular Smad proteins that regulate transcription. Strength and duration of TGFbeta signaling is largely dependent on a negative-feedback program initiated during signal progression. We have identified an inducible gene target of TGFbeta/Smad signaling, the salt-inducible kinase (SIK), which negatively regulates signaling together with Smad7. SIK and Smad7 form a complex and cooperate to down-regulate the activated type I receptor ALK5. We further show that both the kinase and ubiquitin-associated domain of SIK are required for proper ALK5 degradation, with ubiquitin functioning to enhance SIK-mediated receptor degradation. Loss of endogenous SIK results in enhanced gene responses of the fibrotic and cytostatic programs of TGFbeta. We thus identify in SIK a negative regulator that controls TGFbeta receptor turnover and physiological signaling.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I+II antibody produced in mouse, clone JDR.3B8, ascites fluid
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.