MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Oxygenation and adenosine deaminase support growth and proliferation of
Oxygenation and adenosine deaminase support growth and proliferation of
Development (Cambridge, England) (2017-05-21)
Katrin Strassburger, Franziska K Lorbeer, Marilena Lutz, Fabian Graf, Michael Boutros, Aurelio A Teleman
PMID
28526754
ABSTRACT
The
Product Description
H5142
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder
Pricing
S3652
Sigma-Aldrich
Shields and Sang M3 Insect Medium, With L-glutamine and potassium bicarbonate., liquid, sterile-filtered, suitable for insect cell culture
Pricing