  • Search Results
  • Coculture of endothelial progenitor cells and mesenchymal stem cells enhanced their proliferation and angiogenesis through PDGF and Notch signaling.
Coculture of endothelial progenitor cells and mesenchymal stem cells enhanced their proliferation and angiogenesis through PDGF and Notch signaling.
FEBS open bio (2017-11-11)
Tangzhao Liang, Lei Zhu, Wenling Gao, Ming Gong, Jianhua Ren, Hui Yao, Kun Wang, Dehai Shi
ABSTRACT
The beneficial effects of combined use of mesenchymal stem cells (MSCs) and endothelial progenitor cells (EPCs) on tissue repair and regeneration after injury have been demonstrated, but the underlying mechanism remains incompletely understood. This study aimed to investigate the effects of direct contact coculture of human bone marrow-derived EPCs (hEPCs)/human bone marrow-derived MSCs (hMSCs) on their proliferation and angiogenic capacities and the underlying mechanism. hEPCs and hMSCs were cocultured in a 2D mixed monolayer or a 3D transwell membrane cell-to-cell coculture system. Cell proliferation was determined by Cell Counting Kit-8. Angiogenic capacity was evaluated by
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.)
Sigma-Aldrich
Pyridoxine, ≥98%