Studies on Core-Shell Nanocapsules of Felodipine: In Vitro-In Vivo Evaluations.
AAPS PharmSciTech (2017-04-21)
Jerome K Geroge, Priya Ranjan Prasad Verma, Jayachandran Venkatesan, Jin-Young Lee, Dong-Han Yoon, Se-Kwon Kim, Sandeep Kumar Singh
RESUMEN
The present study aimed for in vitro-in vivo-in silico simulation studies of experimentally designed (3
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
[2-(Methacryloyloxy)ethyl]trimethylammonium chloride solution, 75 wt. % in H2O
Tricaprylin, European Pharmacopoeia (EP) Reference Standard