Merck
All Photos(4)

274704

Sigma-Aldrich

Perfluorinated resin solution containing Nafion 1100W

greener alternative

5 wt. % in lower aliphatic alcohols and water, contains 15-20% water

Synonym(s):
Nafion 1100EW
CAS Number:
eCl@ss:
32110203
NACRES:
NA.23

form

liquid

Quality Level

eq. wt.

1,100

contains

15-20% water

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

concentration

5 wt. % in lower aliphatic alcohols and water

refractive index

n20/D 1.379

viscosity

8 cP(20 °C)

density

0.874 g/mL at 25 °C

greener alternative category

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Nafion perfluorinated resin solution is widely used as an electrolytic material. It can be used in the formation of a composite membrane with metal oxides. The membrane further facilitates the formation of chemically stable proton exchange membrane (PEM) fuel cells. It may be coated on mesoporous metallic films to improve the detection and selectivity of biomarkers. It may also form a composite electrode material with graphene for glucose biosensors.
Nafion was used to modify membranes in the study of transport phenomena. Nafion solution was used as an electrolyte in studying the catalytic activity of PtIr electrocatalysts. The suspension was also used in the preparation of Aldose dehydrogenase (ALDH) bioanode. It was used in another study as a component of electrolyte in investigation of mechanico-electrochemical failure modes of the Fe interconnects. 3
Support material for an integrated system which used solar energy to drive chemical reactions. Acts as a superacid catalyst for organic synthesis. When formed as metal composites, can be used as artificial muscle actuators, solid electrolytes for the analysis of electroactive species in organic media, or as electrodes in organic electrosynthesis.

Packaging

25, 100, 500 mL in glass bottle
Packaged in glass bottles

Physical form

Hydrogen ion form

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

67.0 °F

Flash Point(C)

19.44 °C

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service