Merck
All Photos(2)

713716

Sigma-Aldrich

Basosiv M050

greener alternative

produced by BASF

Synonym(s):
Magnesium formate
Empirical Formula (Hill Notation):
C2H2MgO4
CAS Number:
Molecular Weight:
114.34
EC Number:
MDL number:
PubChem Substance ID:

grade

produced by BASF

form

crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

surface area

BET surf. area 400-600 m2/g

bulk density

0.30‑0.40 g/cm3

SMILES string

O=CO[Mg]OC=O

InChI

1S/2CH2O2.Mg/c2*2-1-3;/h2*1H,(H,2,3);/q;;+2/p-2

InChI key

GMDNUWQNDQDBNQ-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Legal Information

Basosiv is a trademark of BASF SE

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service