Merck
All Photos(1)

PEM2A1044

Sigma-Aldrich

Xion PEM-Nafion-1000

thickness 10 μm, L × W 10 cm × 10 cm

L × W

10 cm × 10 cm

thickness

10 μm

SMILES string

FC(CC)(OC(F)(C(F)(F)C(F)(F)OC(F)(C(F)(S(O)(O)=O)F)F)F)CC

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

1 of 4

This Item
PEM2B1066PEM2A3066PEM2B0566
L × W

10 cm × 10 cm

L × W

15 cm × 15 cm

L × W

15 cm × 15 cm

L × W

15 cm × 15 cm

thickness

10 cm × 10 cm

thickness

15 cm × 15 cm

thickness

15 cm × 15 cm

thickness

15 cm × 15 cm

Application

The XION Composite Nafion Membrane is a composite PEM. Its high selectivity, high chemical resistance, and high mechanical strength make it ideal for fuel cell applications. It is stable under neutral and acidic conditions.

Legal Information

Product of Xergy
Nafion is a trademark of The Chemours Company FC, LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service