Skip to Content
Merck

99900222

Opentrons Flex HEPA Filter Replacement Kit

new

Opentrons Flex HEPA Filter Replacement Kit

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

packaging

pack of 1 ea

manufacturer/tradename

999-00222
Opentrons Labworks, Inc.

technique(s)

DNA extraction: suitable
Tissue culture media and additive filtration: suitable

General description

The OPENTRONSFLEX HEPA Filter Replacement Kit will help you easily replace your HEPA filter within the module. The Flex HEPA filter is recommended to be replaced every year if the Flex is used heavily and every 3 years if used moderately (6,000 hours).

Legal Information

OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service