Merck
All Photos(1)

Documents

CRM019

Supelco

Trace Metals - Fly Ash 3

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM019-050

technique(s)

AAS: suitable
ICP: suitable

pH

6.64

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
QC1488QC1052CRM014
Supelco

Supelco

CRM019

Trace Metals - Fly Ash 3

Supelco

Supelco

QC1488

Trace Metals 1 - WS

Supelco

Supelco

QC1052

Trace Metals 2-WP

technique(s)

AAS: suitable, ICP: suitable

technique(s)

AAS: suitable, ICP: suitable

technique(s)

AAS: suitable, ICP: suitable

technique(s)

AAS: suitable, ICP: suitable

application(s)

environmental

application(s)

environmental

application(s)

environmental

application(s)

environmental

format

matrix material

format

matrix material

format

matrix material

format

matrix material

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

certificate is enclosed in each package

packaging

pkg of 50 g

packaging

ampule of 20 mL

packaging

ampule of 20 mL

packaging

pkg of 50 g

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Arsenic
    Barium
    Cadmium
    Chromium
    Copper
    Lead
    Magnesium
    Nickel
    Selenium
    Silver
    Vanadium
    Zinc
    See All (12)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Repr. 1A - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service