Merck
All Photos(1)

Documents

CRM052

Supelco

Trace Metals - Loamy Clay 1

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing

NACRES:
NA.24

grade

certified reference material

CofA

certificate is enclosed in each package

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM052-050

technique(s)

AAS: suitable
ICP: suitable

particle size

0.2-1.0 mm

pH

6.95

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CRM046CRM044CRM2003
vibrant-m

CRM052

Trace Metals - Loamy Clay 1

vibrant-m

CRM046

Trace Metals - Clay 1

vibrant-m

CRM044

Trace Metals - Silt Loam

vibrant-m

CRM2003

Trace Metals - Taiwan Clay 1

application(s)

environmental

application(s)

environmental

application(s)

environmental

application(s)

environmental

format

matrix material

format

matrix material

format

matrix material

format

matrix material

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

CofA

certificate is enclosed in each package

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

packaging

pkg of 50 g

packaging

pkg of 50 g

packaging

pkg of 50 g

packaging

pkg of 50 g

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This product was produced and certified from a naturally contaminated matrix and has not been fortified or modified in any way. Analytical data for Certification was obtained using USEPA SW846, 3rd edition methods 3050 (hot block) and 3051(microwave) using nitric acid extraction. Analysis was carried out according to USEPA methods 6010 (ICP-ES), 6020 (ICP-MS) and 7000 (AES).

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Aluminum
    Antimony
    Arsenic
    Barium
    Beryllium
    Boron
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Lithium
    Magnesium
    Manganese
    Mercury
    Molybdenum
    Nickel
    Potassium
    Selenium
    Silicon
    Silver
    Sodium
    Strontium
    Thallium
    Tin
    Titanium
    Vanadium
    Zinc
    See All (30)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 1

1 of 1

Karolina Lewińska et al.
Environmental monitoring and assessment, 191(2), 70-70 (2019-01-16)
Great concern has been recently focused on antimony in the environment due to the potential risks posed by this metalloid to humans. In Poland, the concentrations of Sb in soils have not been well recognized. The aim of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service