Merck
All Photos(1)

Documents

NIST1568B

Rice flour

NIST® SRM® 1568b

Sign Into View Organizational & Contract Pricing

NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

format

matrix material

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
NIST1567BNIST3234NIST1549A
Rice flour NIST® SRM® 1568b

NIST1568B

Rice flour

Wheat flour NIST® SRM® 1567b

NIST1567B

Wheat flour

vibrant-m

NIST3234

Soy flour

vibrant-m

NIST1549A

Whole milk powder

packaging

pkg of 50 g

packaging

pkg of 50 g

packaging

pkg of 50 g

packaging

pkg of 5 pouches x 10 g ea

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

format

matrix material

format

matrix material

format

matrix material

format

matrix material

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

General description

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phosphorous
Trace Elements: Aluminum (Al), Arsenic (As), Bromine (Br), Cadmium (Cd), Calcium (Ca), Chlorine (Cl2), Cobalt (Co), Copper (Cu), Dimethylarsinic acid, Inorganic arsenic (iAs), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Monomethylarsonic acid (MMA), Potassium (K), Rubidium (Rb), Selenium (Se), Sodium (Na), Sulfur (S), Tin (Sn), Zinc (Zn)

Matrix Group: Cereals, Bread, Rice and Maize

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 4

1 of 4

vibrant-m

BCR465

Rice flour (amylose, low level)

Peanut butter NIST® SRM® 2387

NIST2387

Peanut butter

Bovine liver NIST® SRM® 1577c

NIST1577C

Bovine liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service