Merck
All Photos(1)

RPI-AD8-1

rAct d 8 (Actinidia deliciosa allergen 8.0101), Kiwi

new

pkg of 250 μg

grade

analytical standard

assay

>95%

mol wt

Mw 18 kDa

packaging

pkg of 250 μg

manufacturer/tradename

Indoor Biotechnologies

format

neat

storage temp.

−20°C

General description

Source: Expressed in Pichia pastoris
Formulation: 20mM Tris, 0.3M NaCl, pH 8.0. Filtered through 0.22μm filter.
Concentration: See product insert.
Purification: Multi-step chromatographic purification. Purity > 95 % by SDS-PAGE

Notes:
(1) rAct d 8 has a N-terminal 6xHis-tag.
(2) N-glycosylation motifs at positions 79 and 116 were mutated.

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service