Merck
All Photos(1)

RPI-BE1-1

rBer e 1 (Bertholletia excelsaallergen 1.0101)

new

pkg of 250 μg

grade

analytical standard

assay

>95%

mol wt

Mw 16 kDa

packaging

pkg of 250 μg

manufacturer/tradename

Indoor Biotechnologies

format

neat

storage temp.

−20°C

General description

Source: Pichia pastoris
Formulation: Preservative and carrier-free in 1X PBS, pH 7.4. Filtered through 0.22μm filter.
Concentration: See product insert.
Purification: Purified from Pichia pastoris culture by multi-step chromatography. Purity >95 % by SDS-PAGE, under reducing conditions

Notes:
(1) rBer e 1 has a N-terminal 8xHis-tag.
(2) rBer e 1 (aa 23-146) includes propeptides

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service