
Ascentis® Express C18 (2.7 μm) HPLC Column

Synonym(s):
Core-shell (SPP) Fused Core C18 HPLC column
Products
Product Category
Brand
Product Line
Quality Segment
Application
Technique
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2