MY
EN
Applications
Products
Services
Support
Home
Search Results
Nafion
™
perfluorinated membrane
(2)
Nafion
™
perfluorinated membrane
Linear Formula:
(C
7
HF
13
O
5
S . C
2
F
4
)
x
CAS Number:
31175-20-9
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
915769
Nafion
™
perfluorinated membrane, Nafion
™
117, thickness × L × W 0.007 in. × 0.61 m × 2.5 m, sheet
Expand
View Pricing
915270
Nafion
™
perfluorinated membrane, Nafion
™
117, thickness 0.007 in., sheet size 7.5 cm × 13.0 cm
Expand
View Pricing