Merck
  • Calpain-2 triggers prostate cancer metastasis via enhancing CRMP4 promoter methylation through NF-κB/DNMT1 signaling pathway.
Calpain-2 triggers prostate cancer metastasis via enhancing CRMP4 promoter methylation through NF-κB/DNMT1 signaling pathway.
The Prostate (2018-03-31)
Xin Gao, Yun-Hua Mao, Chutian Xiao, Ke Li, Wei Liu, Liao-Yuan Li, Jun Pang
ABSTRACT
Metastasis is the major cause of cancer-specific death in patients with prostate cancer (PCa). We previously reported that collapsing response mediator protein-4 (CRMP4) is a PCa metastasis-suppressor gene and the hypermethylation in CRMP4 promoter is responsible for the transcription repression in metastatic PCa. However, the underlying mechanisms remain unknown. In this study, we aimed to investigate the role of calpain-2 in CRMP4 promoter hypermethylation and its functional modulation in PCa metastasis. Calpain-2 expression in PCa tissues (n = 87) and its specific mechanisms of functional modulation in CRMP4 expression via limited enzymatic cleavage was investigated. We then focused on the cooperative crosstalk of calpain-2 and NF-κB RelA/p65 in CRMP4 promoter methylation for the initiation of PCa metastasis. Statistical differences between groups were determined using a two-tailed Student's t-test. P < 0.05 indicated statistically significant. Calpain-2 was differentially upregulated in metastatic PCa compared with localized PCa. Moreover, calpain-2 cleaved CRMP4 into the N-terminally fragment which promoted migration and invasion in PCa cells via nuclear translocation and activation of E2F1-mediated DNA methyltransferase 1 (DNMT1) expression. NF-κB RelA/p65 recruited DNMT1 to bind to and methylate CRMP4 promoter in which Serine276 phosphorylation of p65 was essential. Furthermore, CRMP4 exhibited anti-metastatic function via inhibiting the expression of VEGFC through Semaphorin3B-Neuropilin2 signaling. Calpain-2 may contribute to the promoter methylation of CRMP4 to repress its transcription, leading to the metastasis of PCa via enhancing VEGFC expression.
Sigma-Aldrich
MISSION® esiRNA, targeting human DPYSL3
Sigma-Aldrich
MISSION® esiRNA, targeting human RELA