- Long-term complications of sulphur mustard poisoning in severely intoxicated Iranian veterans
Long-term complications of sulphur mustard poisoning in severely intoxicated Iranian veterans
Fundamental & Clinical Pharmacology (2005)
Balali-Mood M, et al.