Merck
  • The RNA Methyltransferase Complex of WTAP, METTL3, and METTL14 Regulates Mitotic Clonal Expansion in Adipogenesis.
The RNA Methyltransferase Complex of WTAP, METTL3, and METTL14 Regulates Mitotic Clonal Expansion in Adipogenesis.
Molecular and cellular biology (2018-06-06)
Masatoshi Kobayashi, Mitsuru Ohsugi, Takayoshi Sasako, Motoharu Awazawa, Toshihiro Umehara, Aya Iwane, Naoki Kobayashi, Yukiko Okazaki, Naoto Kubota, Ryo Suzuki, Hironori Waki, Keiko Horiuchi, Takao Hamakubo, Tatsuhiko Kodama, Seiichiro Aoe, Kazuyuki Tobe, Takashi Kadowaki, Kohjiro Ueki
ABSTRACT
Adipocyte differentiation is regulated by various mechanisms, of which mitotic clonal expansion (MCE) is a key step. Although this process is known to be regulated by cell cycle modulators, the precise mechanism remains unclear. N6-Methyladenosine (m6A) posttranscriptional RNA modification, whose methylation and demethylation are performed by respective enzyme molecules, has recently been suggested to be involved in the regulation of adipogenesis. Here, we show that an RNA N6-adenosine methyltransferase complex consisting of Wilms' tumor 1-associating protein (WTAP), methyltransferase like 3 (METTL3), and METTL14 positively controls adipogenesis by promoting cell cycle transition in MCE during adipogenesis. WTAP, coupled with METTL3 and METTL14, is increased and distributed in nucleus by the induction of adipogenesis dependently on RNA in vitro Knockdown of each of these three proteins leads to cell cycle arrest and impaired adipogenesis associated with suppression of cyclin A2 upregulation during MCE, whose knockdown also impairs adipogenesis. Consistent with this, Wtap heterozygous knockout mice are protected from diet-induced obesity with smaller size and number of adipocytes, leading to improved insulin sensitivity. These data provide a mechanism for adipogenesis through the WTAP-METTL3-METTL14 complex and a potential strategy for treatment of obesity and associated disorders.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
MISSION® esiRNA, targeting human WTAP