Merck
  • CTCF regulates positioning of the human cystic fibrosis gene in association with a histone deacetylase.
CTCF regulates positioning of the human cystic fibrosis gene in association with a histone deacetylase.
Genomics data (2015-10-21)
Joscha Muck
ABSTRACT
The nuclear positioning of mammalian genes often correlates with their functional state. For instance, the human cystic fibrosis transmembrane conductance regulator (CFTR) gene associates with the nuclear periphery in its inactive state, but occupies interior positions when active. Treatment with the histone deacetylase inhibitor trichostatin a (TSA) changes the radial positioning of the CFTR gene in HeLa S3 cells. The gene relocates from the nuclear periphery to the nuclear interior. In Calu-3 cells the gene is located in the nuclear interior. To identify potential regulatory elements for the positioning of CFTR, the histone H3 and H4 acetylation patterns of untreated and TSA-treated HeLa S3 and untreated Calu-3 cells were determined by ChIP-chip. Here is a detailed description of the datasets associated with the study by Muck et al. published in the Journal of Cellular Biochemistry in 2012.
Sigma-Aldrich
GenomePlex® Complete Whole Genome Amplification (WGA) Kit, Optimized kit with enzyme for amplifying a variety of DNA including FFPE tissue