Merck
  • A study of residual lesions in horses that recovered from clinical signs of chronic equine dysautonomia.
A study of residual lesions in horses that recovered from clinical signs of chronic equine dysautonomia.
Journal of veterinary internal medicine (2019-07-25)
Elspeth M Milne, R Scott Pirie, Caroline N Hahn, Jorge Del-Pozo, Dawn Drummond, Sharon Moss, Bruce C McGorum
ABSTRACT
Equine dysautonomia (ED) causes degeneration and loss of autonomic neurons. Approximately 50% of chronic cases recover, but it is unclear how they survive neuronal loss. To assess lesions, autonomic neuron numbers, interstitial cells of Cajal (ICC), and neurodegeneration in recovered cases. Thirteen cases (group ED), euthanized 10.3 ± 5.2 (1-16) years from diagnosis and 6 age-matched controls (group C). Prospective, case control; routine post mortem examination, neuron counts in peripheral and enteric ganglia and immunohistochemical assessment of neural networks (Protein gene product [PGP] 9.5), ICC (c-kit), and neurodegeneration (beta-amyloid precursor protein and ubiquitin) in intestine. Postmortem findings in group ED were small intestinal dilation (4/12, 33%) and muscular hypertrophy (4/12, 33%), and gastric mucosal hypertrophy (3/11, 27%) and ulceration (4/11, 36%). Neuron density was lower in group ED (mean 39% lower for cranial cervical ganglion [P < .001], median 44% lower in celiacomesenteric ganglion [P = .01]). In intestine, neuronal depletion was worst in ileum (median 100% lower in submucosal plexus [P < .001], 91% lower in myenteric plexus [P = .004]). Group ED had less PGP 9.5 staining in ileal myenteric plexus (mean 66% lower [P = .04]) and circular muscle (median 75% lower [P = .006]). In ileum, there was less c-kit staining in myenteric plexus (median 57% lower [P = .02]) but not muscularis externa. Beta-amyloid precursor protein and ubiquitin results were not indicitive of neurodegeneration. Intact ICC in muscularis externa might help maintain motility after neuronal loss. Treatment supporting ICC function warrants investigation.