Merck
  • Metal accumulation in riverine macroinvertebrates from a platinum mining region.
Metal accumulation in riverine macroinvertebrates from a platinum mining region.
The Science of the total environment (2019-11-16)
J H Erasmus, W Malherbe, S Zimmermann, A W Lorenz, M Nachev, V Wepener, B Sures, N J Smit
ABSTRACT
South Africa is the world's main supplier of Pt. The Bushveld Igneous Complex in South Africa contains 75% of the world's Pt resources. Mining of this precious metal requires large volumes of water for production and removal of waste products. Most of this wastewater is discharged into river systems. Although the source of contamination with Pt in aquatic systems due to mining activities is known, little to no information is available about the impact of Pt on aquatic organisms. Additionally, other metals are released as byproducts of Pt mining, which might also be discharged into the environment. Therefore, concentrations of Cr, Ni, Cu, Zn, Cd, Pt and Pb were determined in water, sediment and macroinvertebrate samples from a reference site (Site 1), a highly impacted site (Site 2) and a moderately impacted site (Site 3) along the Hex River, South Africa. Aquatic invertebrate families representing different functional feeding groups i.e. scraper-grazers (Lymnaeidae), collector-gatherers (Potamonautidae, Hydropsychidae, Tubificidae and Chironomidae), shredders (Baetidae) and predators (Coenagrionidae and Libellulidae) were studied. In the sediments, the concentrations of Cr and Pt were significantly higher at Site 2 than at Sites 1 and 3, respectively, whereas concentrations of Ni, Cu, Cd, and Pb showed no significant differences between the sites. Depending on the metal, the aquatic invertebrate families showed different grades of bioaccumulation. The results from especially Lymnaeidae, Baetidae, Tubificidae and Chironomidae showed great promise for the use of these taxa for biomonitoring of metal contaminations. The macroinvertebrates accumulated metals associated with Pt mining, with epi-benthic dwelling taxa (Tubificidae) accumulating higher concentrations of Pt and Cr than other families (e.g. Potamonautidae, Coenagrionidae and Lymnaeidae). These results provide valuable information on the behavior of metals related to Pt mining in aquatic ecosystems and therefore can contribute to the risk assessment of these intensive mining activities.
Sigma-Aldrich
4-Nitro-3-(trifluoromethyl)phenol, 99% (GC)