Merck
  • Type I interferon shapes the quantity and quality of the anti-Zika virus antibody response.
Type I interferon shapes the quantity and quality of the anti-Zika virus antibody response.
Clinical & translational immunology (2020-04-30)
Cheryl Yi-Pin Lee, Guillaume Carissimo, Zheyuan Chen, Fok-Moon Lum, Farhana Abu Bakar, Ravisankar Rajarethinam, Teck-Hui Teo, Anthony Torres-Ruesta, Laurent Renia, Lisa Fp Ng
ABSTRACT
Zika virus (ZIKV) is a mosquito-borne flavivirus that re-emerged in 2015. The association between ZIKV and neurological complications initiated the development of relevant animal models to understand the mechanisms underlying ZIKV-induced pathologies. Transient inhibition of the type I interferon (IFN) pathway through the use of an IFNAR1-blocking antibody, MAR1-5A3, could efficiently permit active virus replication in immunocompetent animals. Type I IFN signalling is involved in the regulation of humoral responses, and thus, it is crucial to investigate the potential effects of type I IFN blockade towards B-cell responses. In this study, comparative analysis was conducted using serum samples collected from ZIKV-infected wild-type (WT) animals either administered with or without MAR1-5A3. Serological assays revealed a more robust ZIKV-specific IgG response and subtype switching upon inhibition of type I IFN due to the abundance of antigen availability. This observation was corroborated by an increase in germinal centres, plasma cells and germinal centre B cells. Interestingly, although both groups of animals recognised different B-cell linear epitopes in the E and NS1 regions, there was no difference in neutralising capacity. Further characterisation of these epitopes in the E protein revealed a detrimental role of antibodies that were generated in the absence of type I IFN. This study highlights the role of type I IFN in shaping the anti-ZIKV antibody response to generate beneficial antibodies and will help guide development of better vaccine candidates triggering efficient neutralising antibodies and avoiding detrimental ones.
Sigma-Aldrich
Human IgG ELISA Kit, for serum, plasma
Pricing and availability is not currently available.