MY
EN
Applications
Products
Services
Support
Home
Search Results
Borane-Tetrahydrofuran
Borane-Tetrahydrofuran
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Zaidlewicz M, et al.