MY
EN
Applications
Products
Services
Support
Home
Search Results
Simple and convenient conversion of acridones into 9-unsubstituted acridines via acridanes using borane tetrahydrofuran complex
Simple and convenient conversion of acridones into 9-unsubstituted acridines via acridanes using borane tetrahydrofuran complex
Tetrahedron Letters (2009)
Desbois N, et al.