Merck
  • Affinity purification of human tau proteins and the construction of a sensitive sandwich enzyme-linked immunosorbent assay for human tau detection.
Affinity purification of human tau proteins and the construction of a sensitive sandwich enzyme-linked immunosorbent assay for human tau detection.
Journal of neurochemistry (1992-02-01)
M Mercken, M Vandermeeren, U Lübke, J Six, J Boons, E Vanmechelen, A Van de Voorde, J Gheuens
ABSTRACT
Immunoaffinity chromatography with a monoclonal antibody produced against bovine tau protein was used to purify tau proteins from human brain. Fifty grams of brain tissue yielded approximately 2 mg of pure tau proteins. The affinity-purified human tau was used to produce a high-titered rabbit anti-human tau serum. The monoclonal anti-tau antibody and the polyclonal rabbit anti-tau serum were then used to construct a sandwich enzyme-linked immunosorbent assay for detection of human tau proteins, with a sensitivity of 1 ng/ml.