Merck
  • Synthesis, biological evaluation, and molecular modeling of berberine derivatives as potent acetylcholinesterase inhibitors.
Synthesis, biological evaluation, and molecular modeling of berberine derivatives as potent acetylcholinesterase inhibitors.
Bioorganic & medicinal chemistry (2010-01-09)
Ling Huang, Anding Shi, Feng He, Xingshu Li
ABSTRACT
By targeting the dual active sites of acetylcholinesterase (AChE), a new series of berberine derivatives was designed, synthesized, and evaluated as AChE inhibitors. Most of the derivatives inhibited AChE in the sub-micromolar range. Compound 8c, berberine linked with phenol by a 4-carbon spacer, showed the most potent inhibition of AChE. A kinetic study of AChE and BuChE indicated that a mix-competitive binding mode existed for these berberine derivatives. Molecular modeling studies confirmed that these hybrids target both the catalytic active site (CAS) and the peripheral anionic site (PAS) of AChE. This is the first report where AChE inhibitory activity has been associated with berberine as a lead molecule.