Merck
  • Investigating the Mechanism of Cyclodextrins in the Treatment of Niemann-Pick Disease Type C Using Crosslinked 2-Hydroxypropyl-β-cyclodextrin.
Investigating the Mechanism of Cyclodextrins in the Treatment of Niemann-Pick Disease Type C Using Crosslinked 2-Hydroxypropyl-β-cyclodextrin.
Small (Weinheim an der Bergstrasse, Germany) (2020-10-21)
Dario Carradori, Hsintsung Chen, Beat Werner, Aagam S Shah, Chiara Leonardi, Mattia Usuelli, Raffaele Mezzenga, Frances Platt, Jean-Christophe Leroux
ABSTRACT
Niemann-Pick disease type C (NPC) is a severe disorder that is characterized by intracellular transport abnormalities leading to cytoplasmic accumulation of lipids such as cholesterol and sphingolipids. The compound 2-hydroxypropyl-β-cyclodextrin (HPβCD) has high cholesterol complexation capacity and is currently under clinical investigation for the NPC treatment. However, due to its short blood half-life, high doses are required to produce a therapeutic effect. In this work, stable polymerized HPβCD is generated to investigate their in vitro mechanisms of action and in vivo effects. Crosslinked CDs (8-312 kDa) display a ninefold greater cholesterol complexation capacity than monomeric HPβCD but are taken up to a lower extent, resulting in an overall comparable in vitro effect. In vivo, the 19.3 kDa HPβCD exhibits a longer half-life than the monomeric HPβCD but it does not increase the life span of Npc1 mice, possibly due to reduced brain penetration. This is circumvented by the application of magnetic resonance imaging-guided low intensity-pulsed focused ultrasound (MRIg-FUS), which increases the brain penetration of the CD. In conclusion, stable polymerized HPβCDs can elucidate CDs' mechanism of action while the use of MRIg-FUS warrants further investigation, as it may be key to harnessing CDs full therapeutic potential in the NPC treatment.
Product Description
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)